Tisk…
Anti-NCOR1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Dodavatel: Bioss
BS-0224R-A350
BOSSBS-0224R-A350EA 25430 CZK
BOSSBS-0224R-A350
Anti-NCOR1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Protilátky
Nuclear co-repressor 2 (N-CoR2) gene (NCOR2, previously called silencing mediator for retinoid and thyroid hormone receptor SMRT) is recruited to nuclear and non-nuclear receptors in a large repressing complex containing also N-CoR1, mSin3 and HDACs. This large complex represses transcription in absence of ligand. Mediates the transcriptional repression activity of some nuclear receptors by promoting chromatin condensation, thus preventing access of the basal transcription. Tissue specificity: Ubiquitous. It is belongs to the N-CoR nuclear receptor corepressors family.
RecommendedMore Product Information