Tisk…

Anti-BACE Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Dodavatel: Bioss
BS-0164R-A488
BOSSBS-0164R-A488EA 25660 CZK
BOSSBS-0164R-A488
Anti-BACE Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Protilátky
Responsible for the proteolytic processing of the amyloid precursor protein (APP). Cleaves at the N-terminus of the A-beta peptide sequence, between residues 671 and 672 of APP, leads to the generation and extracellular release of beta-cleaved soluble APP, and a corresponding cell-associated C-terminal fragment which is later released by gamma-secretase.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BACE
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR