Tisk…

Anti-APAF1 Rabbit Polyclonal Antibody (Cy5®)

Dodavatel: Bioss
BS-0058R-CY5
BOSSBS-0058R-CY5EA 25150 CZK
BOSSBS-0058R-CY5
Anti-APAF1 Rabbit Polyclonal Antibody (Cy5®)
Protilátky
Oligomeric Apaf-1 mediates the cytochrome c-dependent autocatalytic activation of pro-caspase-9 (Apaf-3), leading to the activation of caspase-3 and apoptosis. This activation requires ATP. Isoform 6 is less effective in inducing apoptosis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: APAF1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR