Tisk…
Eco stopcock plugs for lateral burettes, with bore and taper 1:10, BOLA
Dodavatel: Bohlender
J1265-10
BOHLJ1265-10EA 425 CZK
BOHLJ1265-10
Eco stopcock plugs for lateral burettes, with bore and taper 1:10, BOLA
Kohoutky
PTFE/PP.
  • Temperature resistance: from –10 to +120 °C
  • Universal chemical resistance
  • Taper: 1:10
With bore, smooth surface, with grip and nut made of PP, washer included.