Tisk…
Eco stopcock plugs, with taper 1:5
Dodavatel: Bohlender
J1225-15 J1225-10 J1205-19 J1205-18 J1205-13
BOHLJ1225-15EA 234 CZK
BOHLJ1225-15 BOHLJ1225-10 BOHLJ1205-19 BOHLJ1205-18 BOHLJ1205-13
Eco stopcock plugs, with taper 1:5
Kohoutky
Made of PFTE, white, with grip and nut of PP, red.
  • Universal chemical resistance
  • Temperature resistance: from –10 to +120 °C
  • Taper: 1:5
  • With or without bore
  • Washer included