Tisk…
PTFE balls, BOLA
Dodavatel: Bohlender
H964-06 H964-18 H964-03 H964-09 H964-15 H964-21
BOHLH964-06EA 3280 CZK
BOHLH964-06 BOHLH964-18 BOHLH964-03 BOHLH964-09 BOHLH964-15 BOHLH964-21
PTFE balls, BOLA
Kuličky Kuličky plastové
Made from solid PTFE, with smooth surface.
For extension of surfaces of e.g. distillation apparatus; as splash guard.