Tisk…
Screw-in tube fittings
Dodavatel: Bohlender
D518-14 D518-26 D518-36 D518-08 D518-24 D518-12 D518-40 D518-52 D518-70 D518-68 D518-62 D518-54 D518-50 D518-42 D518-38 D518-30 D518-28 D518-18 D518-16 D518-06 D518-04 D518-02
BOHLD518-14EA 736 CZK
BOHLD518-14 BOHLD518-26 BOHLD518-36 BOHLD518-08 BOHLD518-24 BOHLD518-12 BOHLD518-40 BOHLD518-52 BOHLD518-70 BOHLD518-68 BOHLD518-62 BOHLD518-54 BOHLD518-50 BOHLD518-42 BOHLD518-38 BOHLD518-30 BOHLD518-28 BOHLD518-18 BOHLD518-16 BOHLD518-06 BOHLD518-04 BOHLD518-02
Screw-in tube fittings
Fitinky a konektory
Straight tube fitting made of PTFE with nuts made of glass-fibre reinforced PTFE and a screw-in thread (either NPT or G).
  • Universal chemical resistance
  • Temperature resistance from −200 up to 250 °C
  • Pressure: 5 bar
  • Suitable for vacuum