Tisk…
Stirring shafts, fan shaped, BOLA
Dodavatel: Bohlender
C382-02 C382-14 C382-08 C382-06
BOHLC382-02EA 7300 CZK
BOHLC382-02 BOHLC382-14 BOHLC382-08 BOHLC382-06
Stirring shafts, fan shaped, BOLA
Míchadla Hřídele míchací
Stirrer shafts made of PTFE-jacketed stainless steel and the fan shaped stirring unit completely made of PTFE.
  • Suitable for stirring in narrow mouth vessels or in vessels with ground joint openings
  • Chemical resistant
  • Temperature resistance from –200 to +250 °C