Tisk…
Magnetic stirring bars, PTFE, tandem, BOLA
Dodavatel: Bohlender
C363-36 C363-30 C363-39 C363-26
BOHLC363-36EA 4510 CZK
BOHLC363-36 BOHLC363-30 BOHLC363-39 BOHLC363-26
Magnetic stirring bars, PTFE, tandem, BOLA
Míchadla Míchadla magnetická
Tandem shaped magnetic stirrer bars with magnetic core of Alnico 5 encapsulated in PTFE. The centre bore structure is used for receiving bearing neck or a glass neck. In these stirrer bars reduction of running surface to a ring minimises friction and increases lifespan.
  • Chemical resistant
  • Extremely strong mixing can be achieved
  • Ideal transmission of the magnetic force of the stirrer to the tandem magnetic stirring bar
Temperature resistance: −200 to +250 °C.