Tisk…
Sample tubes for stool, with screw cap
Dodavatel: BOETTGER PAUL
07.023.2007 07.043.2007 07.043.3107 07.033.2007 07.023.3107 07.033.3107
BOET07.023.2007EA 9660 CZK
BOET07.023.2007 BOET07.043.2007 BOET07.043.3107 BOET07.033.2007 BOET07.023.3107 BOET07.033.3107
Sample tubes for stool, with screw cap
Systémy pro klinickou diagnostiku Clinical Sample Collection/Preparation Faeces Sample Collection
PP, translucent.
  • Excellent chemical resistance
  • Excellent temperature resistance
Clair tube with overlapping screw cap for optimal hygiene when opening, reliably sealed with long-lasting spoon for about bean-sized samples.
Certifikace: Conforms to DIN EN 829 for the dispatch of stool samples.
Upozornení: *Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904