Tisk…
Microplate shakers, Jitterbug
Dodavatel: BOEKEL SCIENTIFIC
270400 270440
BOEI270400EA 140020 CZK
BOEI270400 BOEI270440
Microplate shakers, Jitterbug
Třepačky Třepačky pro mikrodestičky
Designed to suitable for two or four standard microplates or strip trays.
  • Calibration 'Mode'
  • Temperature calibration
  • Heated lid to provide greater uniformity
  • Independently set temperature, shaking speed and mixing duration