Tisk…
Slide hybridisation oven, RapidFISH
Dodavatel: BOEKEL SCIENTIFIC
240200-2CH 240200-2EU 240200-2UK
BOEI240200-2CHEA 115430 CZK
BOEI240200-2CH BOEI240200-2EU BOEI240200-2UK
Slide hybridisation oven, RapidFISH
Sušárny Pece hybridizační
RapidFISH slide hybridisation oven was designed to simplify fluorescent in-situ hybridisations procedures for FFP samples, cell suspensions, and other tissue preparation procedures.
  • Easy to use control system
  • Automatic clamping slide tray system
  • Superior specimen humidification
  • Excellent temperature performance
  • Programmable heating modes
Temperature control and humidity are essential for FISH probes to bind to RNA and DNA. The RapidFISH has a self-clamping slide tray design that maintains the required humidity. Microscope slides in the RapidFISH doMore Product Information