Tisk…
Dressing jars, with lid
Dodavatel: BOCHEM
8581 8584 8585 8587 8588 8693
BOCH8581EA 1120 CZK
BOCH8581 BOCH8584 BOCH8585 BOCH8587 216-0039 BOCH8693
Dressing jars, with lid
Nádoby Nádoby víceúčelové
Jar made of stainless steel, with knob lid.
  • Very good temperature resistance
  • Good chemical resistance
  • Easy to clean