Tisk…
Stand bases
Dodavatel: BOCHEM
5000 5001 5002 5003
BOCH5000EA 4880 CZK
BOCH5000 BOCH5001 BOCH5002 BOCH5003
Stand bases
Stojany laboratorní
Stand bases made of stainless steel 18/10 with expanded, flat and heavy feet for stability.
  • With M 10 threaded hole (off-centre)
  • One adjustable foot
  • Anti-slip rubber protection
Certifikace: DIN 12892