Tisk…
Wire baskets for transport
Dodavatel: BOCHEM
17300 17301 17302 17305 17306 17307 17310 17311 17312
BOCH17300EA 13030 CZK
BOCH17300 BOCH17301 BOCH17302 BOCH17305 BOCH17306 BOCH17307 BOCH17310 BOCH17311 BOCH17312
Wire baskets for transport
Koše a košíky Koše drátěné
Stainless steel 18/10.
  • Three different mesh dimensions
  • Stackable
  • Excellent thermal and chemical resistance
These rectangular baskets are designed for transporting, storing, cleaning and sterilising laboratory utensils.