Tisk…
Anti-CA9 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: CA9/781]
Dodavatel: Biotium
BNUM0781-50
BNUM0781-50EA 11870 CZK
BNUM0781-50
Anti-CA9 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: CA9/781]
Protilátky
Recognizes a glycoprotein of ~200 kDa, identified as carbonic anhydrase IX (CAIX/gp200). Carbonic Anhydrases (CAs) are members of a large family of zinc metallo-enzymes that catalyze the reversible hydration of carbon dioxide. CAs are involved in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption and the formation of aqueous humor, cerebrospinal fluid, saliva and gastric juice. They show extensive diversity in distribution and in their subcellular localization. CA IX is specifically expressed in clear-cell renal carcinomas.