Tisk…
Anti-CD66 molecule Mouse Monoclonal Antibody (Purified) [clone: C66/1009]
Dodavatel: Biotium
BNUB1009-500 BNUB1009-100
BNUB1009-500EA 14540 CZK
BNUB1009-500 BNUB1009-100
Anti-CD66 molecule Mouse Monoclonal Antibody (Purified) [clone: C66/1009]
Protilátky
This antibody recognizes proteins of 80-200 kDa, identified as different members of CEA family. CEA is synthesized during development in the fetal gut and is re-expressed in increased amounts in intestinal carcinomas and several other tumors. This MAb does not react with nonspecific cross-reacting antigen (NCA) and with human polymorphonuclear leucocytes. It shows no reaction with a variety of normal tissues and is suitable for staining of formalin/paraffin tissues. CEA is not found in benign glands, stroma, or malignant prostatic cells. Antibody to CEA is useful in detecting early foci ofMore Product Information
Objednat
Produktová specifikace
Symbol antigenu CD66 molecule
Název antigenu CD66
Environmentally Preferable
Synonyma antigenu CEACAM5|CD66|Biliary Glycoprotein (BGP-1)|Carcinoembryonic Antigen-related Cell Adhesion Molecule 5
Druh protilátky Primární
Klonovalita Monoclonal
Spojení Unconjugated
Klon Clone C66/1009
Reaktivita Human
Hostitel Mouse
Identifikační číslo genu 169238
Izotyp IgG2a kappa
Imunochemie Yes
ImunoFluorescence Yes
Koncentrace 0,2 mg/ml
Skladovací pufr PBS/0,05% BSA/0,05% azide