Tisk…
Anti-FUT3 Mouse Monoclonal Antibody (Purified) [clone: 2-25LE]
Dodavatel: Biotium
BNUB1008-500 BNUB1008-100
BNUB1008-500EA 14540 CZK
BNUB1008-500 BNUB1008-100
Anti-FUT3 Mouse Monoclonal Antibody (Purified) [clone: 2-25LE]
Protilátky
The Lewis histo-blood group system comprises a set of fucosylated glycosphingolipids that are synthesized by exocrine epithelial cells and circulate in body fluids. The glycosphingolipids function in embryogenesis, tissue differentiation, tumor metastasis, inflammation, and bacterial adhesion. They are secondarily absorbed to red blood cells giving rise to their Lewis phenotype. This gene is a member of the fucosyltransferase family, which catalyzes the addition of fucose to precursor polysaccharides in the last step of Lewis antigen biosynthesis. It encodes an enzyme with alpha(1,3)-More Product Information