Tisk…
Anti-MUC2 Mouse Monoclonal Antibody (Purified) [clone: MLP/842]
Dodavatel: Biotium
BNUB0842-500 BNUB0842-100
BNUB0842-500EA 14540 CZK
BNUB0842-500 BNUB0842-100
Anti-MUC2 Mouse Monoclonal Antibody (Purified) [clone: MLP/842]
Protilátky
Recognizes a single glycoprotein of 520 kDa, identified as mucin 2 (MUC2). This MAb shows no cross-reaction with human milk fat globule membranes, MUC1, or MUC3. Mucins are high molecular weight glycoproteins, which constitute the major component of the mucus layer that protects the gastric epithelium.MUC2 is specifically expressed in goblet cells of the small intestine & colon; in about 65% of colonic carcinomas and about 40% of gastric carcinomas. MUC2 is rarely expressed outside of the GI tract with the exceptions of mucinous carcinoma of breast and clear cell-type carcinomas of theMore Product Information
Objednat
Produktová specifikace
Symbol antigenu MUC2
Název antigenu Mucin 2
Environmentally Preferable
Synonyma antigenu Mucin like protein|SMUC|Intestinal mucin-2|Mucin-2|(MLP)
Druh protilátky Primární
Klonovalita Monoclonal
Spojení Unconjugated
Klon Clone MLP/842
Reaktivita Human
Hostitel Mouse
Identifikační číslo genu 315
Izotyp IgG1 kappa
Imunochemie Yes
ImunoFluorescence Yes
Koncentrace 0,2 mg/ml
Skladovací pufr PBS/0,05% BSA/0,05% azide