Tisk…
Anti-UCHL1 Mouse Monoclonal Antibody (Purified) [clone: UCHL1/841]
Dodavatel: Biotium
BNUB0841-500 BNUB0841-100
BNUB0841-500EA 14540 CZK
BNUB0841-500 BNUB0841-100
Anti-UCHL1 Mouse Monoclonal Antibody (Purified) [clone: UCHL1/841]
Protilátky
This MAb reacts with a protein of 20-30 kDa, identified as PGP9.5, also known as ubiquitin carboxyl-terminal hydrolase-1 (UchL1). Initially, PGP9.5 expression in normal tissues was reported in neurons and neuroendocrine cells but later it was found in distal renal tubular epithelium, spermatogonia, Leydig cells, oocytes, melanocytes, prostatic secretory epithelium, ejaculatory duct cells, epididymis, mammary epithelial cells, Merkel cells, and dermal fibroblasts. Furthermore, immunostaining for PGP9.5 has been shown in a wide variety of mesenchymal neoplasms as well. A mutation in PGP9.5More Product Information