Tisk…
Anti-NKX2-1 Mouse Monoclonal Antibody (Purified) [clone: 8G7G3/1]
Dodavatel: Biotium
BNUB0448-500 BNUB0448-100
BNUB0448-500EA 14540 CZK
BNUB0448-500 BNUB0448-100
Anti-NKX2-1 Mouse Monoclonal Antibody (Purified) [clone: 8G7G3/1]
Protilátky
Recognizes a protein of 40 kDa, identified as Thyroid transcription factor-1 (TTF-1). TTF-1 is a member of the NKx2 family of homeodomain transcription factors. It is expressed in epithelial cells of the thyroid gland and the lung. Nuclei from liver, stomach, pancreas, small intestine, colon, kidney, breast, skin, testes, pituitary, prostate, and adrenal glands are unreactive. Anti-TTF-1 is useful in differentiating primary adenocarcinoma of the lung from metastatic carcinomas originating in the breast, mediastinal germ cell tumors, and malignant mesothelioma. It can also be used toMore Product Information
Objednat
Produktová specifikace
Symbol antigenu NKX2-1
Environmentally Preferable
Název antigenu NK2 homeobox 1
Druh protilátky Primární
Klonovalita Monoclonal
Spojení Unconjugated
Klon Clone 8G7G3/1
Reaktivita Human,Rat,Mouse
Hostitel Mouse
Identifikační číslo genu 94367
Izotyp IgG1 kappa
Imunochemie Yes
ImunoFluorescence Yes
Koncentrace 0,2 mg/ml
Skladovací pufr PBS/0,05% BSA/0,05% azide