Tisk…
Anti-NEFH & NEFL Mouse Monoclonal Antibody (Biotin) [clone: RT-97 NR-4]
Dodavatel: Biotium
BNCB1201-500 BNCB1201-100
BNCB1201-500EA 15940 CZK
BNCB1201-500 BNCB1201-100
Anti-NEFH & NEFL Mouse Monoclonal Antibody (Biotin) [clone: RT-97 NR-4]
Protilátky
This MAb reacts with a 200 kDa and 68 kDa protein, identified as heavy and light sub-units of neurofilaments (NF-H & NF-L). Neurofilaments make up the main structural elements of axons and dendrites and are found in neurons, peripheral nerves, and sympathetic ganglion cells. Neurofilaments consist of three major subunits with molecular weights of 68 kDa (NF-L), 160 kDa (NF-M) and 200 kDa (NF-H). Anti-neurofilament stains a number of neural, neuroendocrine, and endocrine tumors. Neuromas, ganglioneuromas, gangliogliomas, ganglioneuroblastomas, and neuroblastomas stain positively for anti-More Product Information