Tisk…

Anti-CGA Mouse Monoclonal Antibody (CF594) [clone: HCGa/53]

Dodavatel: Biotium
BNC940053-500 BNC940053-100
BNC940053-500EA 15940 CZK
BNC940053-500 BNC940053-100
Anti-CGA Mouse Monoclonal Antibody (CF594) [clone: HCGa/53]
Protilátky
This MAb reacts with a protein of ~13 kDa, identified as alpha subunit of HCG. HCG is a glycoprotein, which is secreted in large quantities by normal trophoblasts. It is present only in trace amounts in non-pregnant urine and sera but rises sharply during pregnancy. HCG is composed of two non-identical, non-covalently linked polypeptide chains designated as the alpha and beta subunits. The alpha subunit is identical to that of thyroid stimulating hormone (TSH), follicle stimulating hormone (FSH), and luteinizing hormone (LH).

Type: Primary
Antigen: CGA
Clonality: Monoclonal
Clone: HCGa/53
More Product Information
Objednat

Produktová specifikace

Symbol antigenu CGA
Environmentally Preferable
Název antigenu Glycoprotein hormones, alpha polypeptide
Druh protilátky Primární
Klonovalita Monoclonal
Spojení CF® 594
Klon Clone HCGa/53
Reaktivita Human
Hostitel Mouse
Identifikační číslo genu 119689
Izotyp IgG1 kappa
Imunochemie Yes
Imunoprecipitace Yes
ImunoFluorescence Yes
Koncentrace 0,1 mg/ml
Skladovací pufr PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR