Tisk…
Anti-ACP5 Mouse Monoclonal Antibody (CF488A) [clone: ACP5/1070]
Dodavatel: Biotium
BNC881070-500 BNC881070-100
BNC881070-500EA 15940 CZK
BNC881070-500 BNC881070-100
Anti-ACP5 Mouse Monoclonal Antibody (CF488A) [clone: ACP5/1070]
Protilátky
This antibody recognizes a protein of 35 kDa, which is identified as tartrate-resistant acid phosphatase (TRAcP). It exists as two isoforms (5a and 5b). This MAb reacts with both the isoforms. Serum TRAcP 5a is secreted by macrophages and dendritic cells and increased in many patients of rheumatoid arthritis.Serum TRAcP 5b is produced from osteoclasts and elevated during bone resorption. TRAcP is an iron containing glycoprotein, which catalyzes the conversion of orthophosphoric monoester to alcohol and orthophosphate. It is the most basic of the acid phosphatases and is the only form notMore Product Information
Objednat
Produktová specifikace
Symbol antigenu ACP5
Environmentally Preferable
Název antigenu TRAcP
Druh protilátky Primární
Klonovalita Monoclonal
Spojení CF® 488A
Klon Clone ACP5/1070
Reaktivita Human,Rat,Mouse
Hostitel Mouse
Identifikační číslo genu 1211
Izotyp IgG2b kappa
Imunochemie Yes
ImunoFluorescence Yes
Koncentrace 0,1 mg/ml
Skladovací pufr PBS/0,1% BSA/0,05% azide