Tisk…
Anti-PAX6 Mouse Monoclonal Antibody (CF488A) [clone: PAX6/498]
Dodavatel: Biotium
BNC880498-500 BNC880498-100
BNC880498-500EA 15940 CZK
BNC880498-500 BNC880498-100
Anti-PAX6 Mouse Monoclonal Antibody (CF488A) [clone: PAX6/498]
Protilátky
Pax genes contain paired domains with strong homology to genes in Drosophila, which are involved in programming early development. Lesions in the Pax-6 gene account for most cases of aniridia, a congenital malformation of the eye, chiefly characterized by iris hypoplasia, which can cause blindness. Pax-6 is involved in other anterior segment malformations besides aniridia, such as Peters anomaly, a major error in the embryonic development of the eye with corneal clouding with variable iridolenticulocorneal adhesions. The Pax-6 gene encodes a transcriptional regulator that recognizes targetMore Product Information