Tisk…
Anti-TAG-72 / CA72.4 Mouse Monoclonal Antibody (CF568) [clone: B72.3]
Dodavatel: Biotium
BNC680276-500 BNC680276-100
BNC680276-500EA 15940 CZK
BNC680276-500 BNC680276-100
Anti-TAG-72 / CA72.4 Mouse Monoclonal Antibody (CF568) [clone: B72.3]
Protilátky
Recognizes an oncofetal antigen of 220 kDa, identified as a tumor-associated glycoprotein (TAG-72) with properties of a mucin. This MAb defines the mucin-carried sialylated-Tn epitope. TAG-72 is usually expressed by adenocarcinomas, but is negative in mesotheliomas. Studies have reported that this antibody has 80% sensitivity and 93% specificity for pulmonary adenocarcinoma. Therefore, TAG-72 is a useful marker to distinguish between mesothelioma and adenocarcinoma. However, false positive reactions can occur so results must be interpreted with the utmost caution. This antibody may be usefulMore Product Information
Objednat
Produktová specifikace
Environmentally Preferable
Název antigenu TAG-72 / CA72.4
Synonyma antigenu CA72.4|Tumor associated glycoprotein 72
Druh protilátky Primární
Klonovalita Monoclonal
Spojení CF® 568
Klon Clone B72.3
Reaktivita Hamster,Human,Bovine,Dog,Rat
Hostitel Mouse
Izotyp IgG1 kappa
ImunoFluorescence Yes
Koncentrace 0,1 mg/ml
Skladovací pufr PBS/0,1% BSA/0,05% azide