Tisk…
Anti-CD66 molecule Mouse Monoclonal Antibody (CF647) [clone: C66/195 C66/261 C66/1009 C66/1030]
Dodavatel: Biotium
BNC471139-500 BNC471139-100
BNC471139-500EA 15940 CZK
BNC471139-500 BNC471139-100
Anti-CD66 molecule Mouse Monoclonal Antibody (CF647) [clone: C66/195 C66/261 C66/1009 C66/1030]
Protilátky
This antibody recognizes proteins of 80-200 kDa, identified as different members of CEA family. CEA is synthesized during development in the fetal gut and is re-expressed in increased amounts in intestinal carcinomas and several other tumors. This MAb does not react with nonspecific cross-reacting antigen (NCA) and with human polymorphonuclear leucocytes. It shows no reaction with a variety of normal tissues and is suitable for staining of formalin/paraffin tissues. CEA is not found in benign glands, stroma, or malignant prostatic cells. Antibody to CEA is useful in detecting early foci ofMore Product Information