Tisk…
Anti-Human Nuclear Antigen Mouse Monoclonal Antibody (CF543) [clone: 235-1]
Dodavatel: Biotium
BNC430099-500 BNC430099-100
BNC430099-500EA 15940 CZK
BNC430099-500 BNC430099-100
Anti-Human Nuclear Antigen Mouse Monoclonal Antibody (CF543) [clone: 235-1]
Protilátky
This MAb is an excellent marker for human cells in xenographic model research. It reacts specifically with human cells. It is a part of a new panel of reagents, which recognizes subcellular organelles or compartments of human cells. These markers may be useful in identification of these organelles in cells, tissues, and biochemical preparations. MAb 235-1 recognizes an antigen associated with the nuclei in human cells. It can be used to stain the nuclei in cell or tissue preparations and can be used as a nuclear marker in subcellular fractions. It produces a speckled pattern in normal andMore Product Information