Tisk…
Anti-CGB Mouse Monoclonal Antibody (CF405S) [clone: HCGb/211]
Dodavatel: Biotium
BNC040459-500 BNC040459-100
BNC040459-500EA 15940 CZK
BNC040459-500 BNC040459-100
Anti-CGB Mouse Monoclonal Antibody (CF405S) [clone: HCGb/211]
Protilátky
This MAb reacts with a protein of 22 kDa, identified as beta subunit of HCG. It does not cross react with the alpha subunit. HCG is a glycoprotein, which is secreted in large quantities by normal trophoblasts. It is present only in trace amounts in non-pregnant urine and sera but rises sharply during pregnancy. HCG is composed of two non-identical, non-covalently linked polypeptide chains designated as the alpha and beta subunits. The alpha subunit is identical to that of thyroid stimulating hormone (TSH), follicle stimulating hormone (FSH), and luteinizing hormone (LH). hCG MAb detectsMore Product Information