Tisk…
Anti-Small Cell Lung Cancer Mouse Monoclonal Antibody (CF405S) [clone: MOC-52]
Dodavatel: Biotium
BNC040329-500 BNC040329-100
BNC040329-500EA 15940 CZK
BNC040329-500 BNC040329-100
Anti-Small Cell Lung Cancer Mouse Monoclonal Antibody (CF405S) [clone: MOC-52]
Protilátky
This MAb reacts with a membrane-associated protein present in normal and malignant neuroendocrine tissues including small cell lung cancer (SCLC). It stains neural and a variable number of endocrine tissues and in the lung it reacts preferentially with SCLC and carcinoids. Its epitope is destroyed during formalin fixation. This antibody was categorized during the First International Workshop on Small Cell Lung Cancer Antigens held in London in April 1987. There are two major types of Lung Carcinoma: non-small cell, which accounts for 80% of all cases; and small cell, which accounts forMore Product Information