Tisk…
Anti-HIF1A Mouse Monoclonal Antibody (CF405S) [clone: HIF1A-84]
Dodavatel: Biotium
BNC040084-500 BNC040084-100
BNC040084-500EA 15940 CZK
BNC040084-500 BNC040084-100
Anti-HIF1A Mouse Monoclonal Antibody (CF405S) [clone: HIF1A-84]
Protilátky
HIF1 (hypoxia-inducible factor 1), a heterodimeric transcription factor complex central to cellular response to hypoxia, consists of two subunits (HIF-1 alpha and HIF-1 beta) which are basic helix-loop-helix proteins of the PAS (Per, ARNT, Sim) family. Expression of HIF-1 alpha protein is regulated by cellular oxygen level alterations as well as in oxygen-independent manner via different cytokines (through the PI3K-AKT-mTOR pathway), growth factors, oncogenic activation, or loss of tumor suppressor function etc. In normoxic cells, HIF-1 alpha is proline hydroxylated leading to aMore Product Information
Objednat
Produktová specifikace
Symbol antigenu HIF1A
Environmentally Preferable
Název antigenu HIF1-alpha
Druh protilátky Primární
Klonovalita Monoclonal
Spojení CF® 405S
Klon Clone HIF1A-84
Reaktivita Human
Hostitel Mouse
Identifikační číslo genu 597216
Izotyp IgG2b kappa
ImunoFluorescence Yes
Koncentrace 0,1 mg/ml
Skladovací pufr PBS/0,1% BSA/0,05% azide