Tisk…

Anti-GLI2 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB97067-100
BIRBORB97067-100EA 0 CZK
BIRBORB97067-100
Anti-GLI2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: GLI2 (GLI family zinc finger 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR