Tisk…

Anti-OVCA2 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB76752-50
BIRBORB76752-50EA 0 CZK
BIRBORB76752-50
Anti-OVCA2 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:1000

Type: Primary
Antigen: OVCA2 (ovarian tumor suppressor candidate 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR