Tisk…

Anti-ITGA3 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB76135-100
BIRBORB76135-100EA 0 CZK
BIRBORB76135-100
Anti-ITGA3 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ITGA3 (integrin, alpha 3 (antigen CD49C, alpha 3 subunit of VLA-3 receptor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Objednat

Produktová specifikace

Symbol antigenu ITGA3
Environmentally Preferable
Název antigenu Integrin Alpha 3
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Unconjugated
Reaktivita
Hostitel Rabbit
Izotyp IgG
Western blot Yes
Imunochemie Yes
Imunogen A synthetic peptide corresponding to a sequence at the N-terminus of human ITGA3(51-67aa LFGYSVALHRQTERQQR), identical to the related rat and mouse sequences.
Počet aminokyselin 51 to 67
Sekvence LFGYSVALHRQTERQQR
Formulář lyophilized
Čištění purified by immunoaffinity chromatography
Koncentrace 0,1-0,5 ug/ml,0,5-1 ug/ul
Skladovací pufr 5mg BSA, 0,9mg NaCl, 0,2mg Na2HPO4, 0,05mg Thimerosal, 0,05mg NaN3.
Skladovací teplota Cool
Transportní teplota Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR