Tisk…

Anti-SARS Spike Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB74525-100
BIRBORB74525-100EA 0 CZK
BIRBORB74525-100
Anti-SARS Spike Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SARS Spike
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Virus
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR