Tisk…

Anti-EDA Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB48027-100
BIRBORB48027-100EA 0 CZK
BIRBORB48027-100
Anti-EDA Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: EDA (ectodysplasin A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR