Tisk…

Anti-Myc Tag Mouse Monoclonal Antibody [clone: Myc.A+]

Dodavatel: Biorbyt
ORB22963-100
BIRBORB22963-100EA 0 CZK
BIRBORB22963-100
Anti-Myc Tag Mouse Monoclonal Antibody [clone: Myc.A+]
Protilátky
Type: Primary
Antigen: Myc Tag (Myc tag)
Clonality: monoclonal
Clone: Myc.A+
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR