Tisk…

Anti-LAT Goat Polyclonal Antibody

Dodavatel: Biorbyt
ORB20330-100
BIRBORB20330-100EA 0 CZK
BIRBORB20330-100
Anti-LAT Goat Polyclonal Antibody
Protilátky
ELISA: 1:64000; Western Blot: 1-3 ?g/ml

Type: Primary
Antigen: LAT (linker for activation of T cells)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR