Tisk…

Anti-KL Goat Polyclonal Antibody

Dodavatel: Biorbyt
ORB20197-100
BIRBORB20197-100EA 0 CZK
BIRBORB20197-100
Anti-KL Goat Polyclonal Antibody
Protilátky
ELISA: 1:1000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: KL (klotho)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR