Tisk…

Anti-EPEC Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Biorbyt
ORB17007-100 ORB17007-200
BIRBORB17007-100EA 0 CZK
BIRBORB17007-100 BIRBORB17007-200
Anti-EPEC Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: EPEC
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR