Tisk…

Anti-KL Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy5®)

Dodavatel: Biorbyt
ORB130000-100
BIRBORB130000-100EA 0 CZK
BIRBORB130000-100
Anti-KL Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy5®)
Protilátky
Type: Primary
Antigen: KL (klotho)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR