Tisk…

Anti-PPP1R8 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB128562-100 ORB128562-200
BIRBORB128562-100EA 0 CZK
BIRBORB128562-100 BIRBORB128562-200
Anti-PPP1R8 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PPP1R8 (protein phosphatase 1, regulatory subunit 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR