Tisk…

Anti-ACAD10 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB125378-100 ORB125378-200
BIRBORB125378-100EA 0 CZK
BIRBORB125378-100 BIRBORB125378-200
Anti-ACAD10 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ACAD10 (acyl-CoA dehydrogenase family, member 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR