Tisk…

Anti-APOC4 Rabbit polyclonal antibody

Dodavatel: Biorbyt
ORB101551-100 ORB101551-200
BIRBORB101551-100EA 0 CZK
BIRBORB101551-100 BIRBORB101551-200
Anti-APOC4 Rabbit polyclonal antibody
Protilátky
  • ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: APOC4 (apolipoprotein C-IV)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR