Tisk…
BioSample cards
Dodavatel: Ahlstrom-Munksjö
8.460.0001.B-N 8.460.0012.A-N 8.460.0004.A-N 8.460.0004.B-N 8.460.0005.B-N 8.460.0038.A-N 8.294.0001.B-N 2.460.210297
BINZ8.460.0001.B-NEA 6080 CZK
BINZ8.460.0001.B-N BINZ8.460.0012.A-N BINZ8.460.0004.A-N BINZ8.460.0004.B-N BINZ8.460.0005.B-N BINZ8.460.0038.A-N BINZ8.294.0001.B-N BINZ2.460.210297
BioSample cards
Systémy pro klinickou diagnostiku Clinical Sample Collection/Preparation Biological Sample Collection/Preparation
These BioSample cards are designed for microvolume sampling, transport and ambient storage of biological samples. These cards are made of white and pure absorbent fibers and contain no wet-strength additives or chemicals. This feature ensures reproducible collection and minimise possible interferences for reliable analysis of targeted analytes like small molecules, proteins, nucleic acids or vitamins.
  • Reliable and reproducible sample collection
  • Even and uniform sample distribution
  • Customisable design and printing
  • Rigorous quality control
  • Each card has five/
More Product Information