Tisk…
Filter paper for special applications
Dodavatel: Ahlstrom-Munksjö
146156 146158 146162 146166 146164 146191 146322 146174 146324 146160 146320 146339 146337 146336 146333 146332 146330 146327 144032 144043 146172 146200 146201 146154 146168 146169 146175 146177 146187 146188 146190 146340 146341
BINZ146156EA 475 CZK
BINZ146156 BINZ146158 BINZ146162 BINZ146166 BINZ146164 BINZ146191 BINZ146322 BINZ146174 BINZ146324 BINZ146160 BINZ146320 BINZ146339 BINZ146337 BINZ146336 BINZ146333 BINZ146332 BINZ146330 BINZ146327 BINZ144032 BINZ144043 BINZ146172 BINZ146200 BINZ146201 BINZ146154 BINZ146168 BINZ146169 BINZ146175 BINZ146177 BINZ146187 BINZ146188 BINZ146190 BINZ146340 BINZ146341
Filter paper for special applications
Filtry Papíry filtrační Papír kvalitativní filtrační
These filter papers for suitable for special applications.
Grade 11: Plain, hardened.
Grade 389F: Plain, for determination of fat in foodstuffs.
Grade 480: Plain, phase separation paper, hydrophobic silicone-impregnated paper.
Grade 918: Black filter paper, stained with a sulfur colouring to reveal the particles of bright colour, detection of fluorine or silicon and mycelium in cultivated mushrooms.
Grade 470: Plain, kieselguhr (diatomite) paper for fine turbidity.
Grade 69/K: Plain, activated carbon, used in laboratory: clarification and brightening of dull and dark urines,More Product Information