Tisk…
Nessler cylinders
Dodavatel: SciLabware
CRC320 CRC322
BIBBCRC320EA 794 CZK
BIBBCRC320 BIBBCRC322
Nessler cylinders
Zkumavky Zkumavky dle Nesslera
These Nessler cylinders are designed for colour comparison of water samples, foodstuffs, etc
  • Made of specially selected borosilicate glass tubing
  • Supplied in matched pairs for uniformity