Tisk…
Kjeldahl digestion and distillation assembly, Quickfit®
Dodavatel: SciLabware
280MC
BIBB280MCEA 12440 CZK
BIBB280MC
Kjeldahl digestion and distillation assembly, Quickfit®
Systémy rozkladové Mineralizátory
Convenient assembly of Quickfit® glassware for Kjeldahl digestion (nitrogen determination) and distillation.
  • Manufactured from heat resistant and chemically durable borosilicate glass
Flask can be used alone for digestion stage and then fitted to rest of assembly for the distillation process.