Tisk…
Cleanroom notebooks, BCR® MyNotes
Dodavatel: BERKSHIRE
MNBK-A5.CR MNBK-A4.CR MNBKMEMOCR MNBKA5GRID MNBKA5CR
BERKMNBK-A5.CREA 522 CZK
BERKMNBK-A5.CR BERKMNBK-A4.CR BERKMNBKMEMOCR BERKMNBKA5GRID BERKMNBKA5CR
Cleanroom notebooks, BCR® MyNotes
Zápisníky
BCR® MyNotes Notebooks are an ideal choice for documentation. They are practical for note taking and data recording in the cleanroom (Class 1 - 10,000).
  • Made with Berkshire cleanroom bond 850 (medium 85 g/m² (24 lbs.))
  • Low in particles and fibres
  • Full lot tracking on every case
  • Double packed for cleanliness
  • Smooth surfaces reduce particle contamination
  • Available in college rule (CR) or 1/4" grid options
BCR® Mynotes are made with the purest quality cellulose from specially selected forests and is specially coated to minimise particle and fiber release.More Product Information