Tisk…
Cleanroom wipes, sterile, Gamma Wipe® 67
Dodavatel: BERKSHIRE
GW67.ST.60 GW67.ST.25 GW67ST0980
115-0262EA 28380 CZK
115-0262 115-0348 BERKGW67ST0980
Cleanroom wipes, sterile, Gamma Wipe® 67
Ubousky/Utěrky Utěrky pro čisté prostory Ubrousky sterilní do čistých prostor
Designed for general sterile cleanroom applications requiring cleanliness, durability, and high absorbency, made of a low-particulating, non woven blend of 45% polyester and 55% cellulose.
  • Certificate of Sterility is included with each case
  • Sterilised by gamma radiation to a Sterility Assurance Level of 10⁻⁶
Suitable for Class 100, ISO 5, M3,5 environments.
Certifikace: According to ANSI/ISO/AAMI guidelines.
Balení: Packed in peel-away header bags.
Upozornení: Data accurate at time of publishing, latest test results and test methods can beMore Product Information